Learn More
Sino Biological Influenza A H5N1 (A/Xinjiang/1/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40004V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 340) of the influenza hemagglutinin (A/Xinjiang/1/2006 (H5N1)) (ACJ68614.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0069