Learn More
Sino Biological Influenza A H1N1 (A/Ohio/07/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40007V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of the influenza A H1N1 (A/Ohio/07/2009 (H1N1)) hemagglutinin (ACQ63286.1) (Met 1-Gln 529, HA1+HA2, uncleved), was fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-0083