Learn More
Sino Biological Influenza A H4N4 (A/mallard duck/Alberta/299/1977) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40008V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard duck/Alberta/299/1977(H4N4)) hemagglutinin (ABB87495.1) (Met1-Ile528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0087