Learn More
Sino Biological Influenza A H1N1 (A/New York/18/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40009V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/New York/18/2009(H1N1)) hemagglutinin (ACR08536.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0093