Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/34/Mount Sinai) Matrix protein 1 / M1 Protein (His Tag)
Supplier:  Sino Biological 40010V07E100
Encompass_Preferred
A DNA sequence encoding the influenza A H1N1 Virus (A/Puerto Rico/8/34/Mount Sinai) M1 protein (NP_040978.1) (Ser 2-Lys 252) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0101