Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/34/Mount Sinai) Non-structural / NS1 Protein (His Tag)
Supplier:  Sino Biological 40011V07E100
Encompass_Preferred
A DNA sequence encoding the influenza A H1N1 Virus (A/Puerto Rico/8/34/Mount Sinai) NS1 protein (C8XP22) (Asp 2-Val 230) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0103