Learn More
Sino Biological Influenza A H5N1 (A/Hubei/1/2010) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40015V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/Hubei/1/2010 (H5N1)) (Met 1-Gln 530) (HA1 + HA2, cleaved) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0114