Learn More
Sino Biological Influenza B (B/Brisbane/60/2008) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40016V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Brisbane/60/2008) hemagglutinin (ACN29380.1) (Met1-Ala556), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0116