Learn More
Sino Biological Influenza B (B/Brisbane/60/2008) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40016V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 556) of the influenza B hemagglutinin (B/Brisbane/60/2008)) (ACN29380.1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0119