Learn More
Sino Biological Influenza B (B/Brisbane/60/2008) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40016V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Met1-Arg362) of the influenza B hemagglutinin (influenza B virus (B/Brisbane/60/2008)) (ACN29383.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0121