Learn More
Sino Biological Influenza A H5N1 (A/Vietnam/UT31413II/2008) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40022V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Vietnam/UT31413II/2008)(H5N1)) hemagglutinin (ADF83651.1) (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0148