Learn More
Sino Biological Influenza A H5N1 (A/Vietnam/UT31413II/2008) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40022V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met1-Arg339) of the Influenza A Virus hemagglutinin (A/Vietnam/UT31413II/2008(H5N1)) (ADF83651.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-0149