Learn More
Sino Biological Influenza A H5N1 (A/goose/Guangdong/1/1996) Hemagglutinin Protein (His Tag)
Supplier:  Sino Biological 40024V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/goose/Guangdong/1/1996(H5N1)) hemagglutinin (YP_308669.1) (Met1-Glu340), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0154