Learn More
Sino Biological Influenza A H4N8 (A/chicken/Alabama/1/1975) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40025V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza hemagglutinin (A/chicken/Alabama/1/1975 (H4N8)) (P19695.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0157