Learn More
Sino Biological Influenza A H6N4 (A/chicken/Hong Kong/17/1977) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40027V08H1100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/chicken/Hong Kong/17/1977 (H6N4)) (CAC84244.1) (Met 1-Gln 529), was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0169