Learn More
Sino Biological Influenza A H10N9 (A/duck/Hong Kong/562/1979) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40028V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 340) of the influenza hemagglutinin (A/duck/Hong Kong/562/1979 (H10N9)) (ABI84469.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0177