Learn More
Sino Biological Influenza A H9N2 (A/chicken/Hong Kong/G9/1997) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40034V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/chicken/Hong Kong/G9/1997(H9N2)) neuraminidase (AAD49001.1) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-162-0190