Learn More
Sino Biological Influenza A H1N1 (A/Beijing/22808/2009) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40035V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 344) of the influenza hemagglutinin (A/Beijing/22808/2009 (H1N1)) (ADD64203.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0196