Learn More
Sino Biological Influenza A H9N2 (A/chicken/Hong Kong/G9/1997) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40036V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 338) of the influenza hemagglutinin (Influenza A virus (A/chicken/Hong Kong/G9/1997 (H9N2)) (AAF00701.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0201