Learn More
Sino Biological Human respiratory syncytial virus (RSV) Fusion protein / RSV-F (Strain RSS-2) Protein (His Tag)
Supplier:  Sino Biological 40037V08B100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human RSV (strain RSS-2) fusion protein (P11209) (Met 1-Thr 529) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0205