Learn More
Sino Biological Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40040V07H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/1073/99(H9N2)) neuraminidase (NP_859038.1) (His36-Ile469) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-0207