Learn More
Sino Biological Influenza A H3N2 (A/Perth/16/2009) Hemagglutinin / HA-specific B cell probe (His Tag)
Supplier:  Sino Biological 40043V08B1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Perth/16/2009 (H3N2)) hemagglutinin (ACS71642.1) (Met1-Lys519, with a mutation Tyr 108 Phe), was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0209