Learn More
Sino Biological Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40044V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) hemagglutinin (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0220