Learn More
Sino Biological Influenza A H3N2 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40058V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 345) of theinfluenza hemagglutinin (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)(H3N2))(ADI52838.1), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0233