Learn More
Sino Biological Influenza A H3N2 (A/X-31) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40059V08H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/X-31 (H3N2)) hemagglutinin (P03438) (Met1-Trp530), termed as HA, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0237