Learn More
Sino Biological Recombinant Listeria monocytogenes flagellin Protein (His Tag)
Supplier:  Sino Biological 40067V07E100
Encompass_Preferred
A DNA sequence encoding the Listeria monocytogenes flaA (YP_013330.1) (Met1-Ser287) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-0249