Learn More
Sino Biological MERS-CoV Nucleoprotein / NP Antibody, Rabbit PAb
Supplier:  Sino Biological 40068RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant MERS-CoV (NCoV / Novel coronavirus) Nucleoprotein / NP (Catalog#40068-V08B; AFS88943.1; Met1-Asp413). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-202-2472