Learn More
Sino Biological MERS-CoV Spike Protein (S1+S2 ECD, aa 1-1297, His Tag)
Supplier:  Sino Biological 40069V08B100
Encompass_Preferred
A DNA sequence encoding the MERS-CoV extracellular domain of spike protein (AFS88936.1) (Met1-Trp1297) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0255