Learn More
Sino Biological MERS-CoV Spike/RBD Protein fragment (RBD, aa 367-606, His Tag)
Supplier:  Sino Biological 40071V08B1100
Encompass_Preferred
A DNA sequence encoding the MERS-CoV receptor binding domain (RBD) of spike protein fragment (AFS88936.1) (Glu367-Tyr606) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0275