Learn More
Sino Biological MERS-CoV Spike/RBD Protein fragment (RBD, aa 367-606, rFc Tag)
Supplier:  Sino Biological 40071V31B1100
Encompass_Preferred
A DNA sequence encoding the receptor binding domain (RBD) of MERS-CoV spike protein fragment (AFS88936.1) (Glu367-Tyr606) was fused with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-0280