Learn More
Sino Biological Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP1 (mucin domain deleted) Protein (aa:Met1-Asp320, His Tag)
Supplier:  Sino Biological 40094V08B100
Encompass_Preferred
A DNA sequence encoding Sudan ebolavirus (strain Uganda-00) GP (Q7T9D9.1) (Met1-Asp320) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0297