Learn More
Sino Biological Ebola virus EBOV (subtype Sudan, strain Gulu) GP1 / Glycoprotein Protein (His Tag)
Supplier:  Sino Biological 40094V08H2100
Encompass_Preferred
A DNA sequence encoding the Sudan ebolavirus (strain Gulu) GP (YP_138523.1) (Met1-Arg501) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0307