Learn More
Chemglass Life Sciences 400mL Beaker, Low Form, Griffin, Pyrex
SDP
Catalog No. 501215007
Encompass_Preferred
400mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. The 250 through 2000mL have a double graduated scale to indicate approximate volume. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.
Catalog No. 50-121-5007 Supplier Chemglass Life Sciences Supplier No. CG8048P400
400mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. The 250 through 2000mL have a double graduated scale to indicate approximate volume. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.