Learn More
Ace Glass, Inc. Beaker, 400mL, jacketed, glass hose connections for 3/8intubing, 112mm inside height, 75mm inside diameter
SDP
Catalog No. 50971845
Encompass_Preferred
400mL jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 112mm I.H. x 75mm I.D. Beaker does not have a pouring spout.
Catalog No. 50-971-845 Supplier Ace Glass, Inc. Supplier No. 534010
Add to Cart
Add to Cart
400mL jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 112mm I.H. x 75mm I.D. Beaker does not have a pouring spout.