Learn More
S2 Media 400mL Sabouraud Dextrose Agar with Lecithin and Tween 80 (SabDex w/LT) in 500mL Media Bottle, 1ea
Catalog No. 502995898
Encompass_Preferred
Sabouraud Dextrose Agar (SDA, SabDex) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. Lecithin and Tween 80 are added to neutralize the antimicrobial effects of disinfectants and cleaning solutions used on environmental surfaces.• Product meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• SabDex w/LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5898 Supplier S2 Media Supplier No. 5250
Sabouraud Dextrose Agar (SDA, SabDex) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. Lecithin and Tween 80 are added to neutralize the antimicrobial effects of disinfectants and cleaning solutions used on environmental surfaces.• Product meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• SabDex w/LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.