Learn More
S2 Media 400mL Sabouraud Dextrose Agar in 500mL Media Bottle, 1ea
Catalog No. 502995753
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. • Product meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• SabDex should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5753 Supplier S2 Media Supplier No. 5243
Sabouraud Dextrose Agar (SDA) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. • Product meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• SabDex should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.