Learn More
S2 Media 500mL Tryptic Soy Agar (TSA)/Soybean-Casein Digest (SCD) in 500mL Media Bottle, 1ea
Catalog No. 502995752
Encompass_Preferred
Tryptic Soy Agar (TSA) is a non-selective and non-differential, general growth medium. It is recommended for the isolation and cultivation of fastidious and non-fastidious microorganisms, although it is not the medium of choice for anaerobes. TSA is also recommended for the preparation, maintenance, and storage of test strains. Lecithin and Tween 80 are added to neutralize the antimicrobial effects of disinfectants and cleaning solutions used on environmental surfaces.• Product complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5752 Supplier S2 Media Supplier No. 5257
Tryptic Soy Agar (TSA) is a non-selective and non-differential, general growth medium. It is recommended for the isolation and cultivation of fastidious and non-fastidious microorganisms, although it is not the medium of choice for anaerobes. TSA is also recommended for the preparation, maintenance, and storage of test strains. Lecithin and Tween 80 are added to neutralize the antimicrobial effects of disinfectants and cleaning solutions used on environmental surfaces.• Product complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.