Learn More
Ace Glass, Inc. 400mm x 10mm OD porosity A straight sparger tube, 1/2" connection top
SDP
Catalog No. 50973017
Encompass_Preferred
400mm X 10mm O.D. straight sparger tube with a 145-174 micron fritted disc at the bottom, and a 1/2 I.D. hose connection tubing at the top. Will fit inside #11 Ace-Threds.
Catalog No. 50-973-017 Supplier Ace Glass, Inc. Supplier No. 645304
400mm X 10mm O.D. straight sparger tube with a 145-174 micron fritted disc at the bottom, and a 1/2 I.D. hose connection tubing at the top. Will fit inside #11 Ace-Threds.