Learn More
Aqua Solutions 400PPM TOC STD 1L
SDP
Catalog No. NC0471691
Encompass_Preferred
Total Organic Carbon Standard 400 Ppm Solution, Prepared To Epa 415 Standard Methods (1l)
Catalog No. NC0471691 Supplier Aqua Solutions Supplier No. 95961L
Total Organic Carbon Standard 400 Ppm Solution, Prepared To Epa 415 Standard Methods (1l)