Learn More
Sino Biological Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40103V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Anhui/1/2013(H7N9)) hemagglutinin (translated amino acid sequence of EPI439507) (Met1-Val524) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0312