Learn More
Sino Biological Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40104V08B1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Shanghai/1/2013(H7N9)) hemagglutinin (Met1-Gly338), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0326