Learn More
Sino Biological Influenza A H7N9 (A/Hangzhou/1/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40105V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hangzhou/1/2013 (H7N9)) (AGI60301.1) hemagglutinin (Met1-Val524) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0341