Learn More
Sino Biological Influenza A H7N9 (A/Pigeon/Shanghai/S1069/2013) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40106V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Pigeon/Shanghai/S1069/2013(A/H7N9)) hemagglutinin (translated amino acid of EPI440701) (Met1-Val524) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0347