Learn More
Sino Biological Influenza A H2N2 (A/Guiyang/1/1957) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40119V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Guiyang/1/1957(H2N2)) hemagglutinin (ACD85231.1) (Met1-Arg340), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0394