Learn More
Sino Biological Influenza A H7N3 (A/turkey/Italy/214845/2002) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40128V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/turkey/Italy/214845/2002(H7N3)) hemagglutinin (CAF33017.1) (Met1-Arg349), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0409