Learn More
Sino Biological Influenza A H1N1 (A/Texas/36/1991) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40132V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Texas/36/1991(H1N1)) hemagglutinin (ACF41933.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0417