Learn More
Sino Biological Influenza A H1N1 (A/USSR/90/1977) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40134V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/USSR/90/1977(H1N1)) hemagglutinin (P03453) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0425