Learn More
Sino Biological Influenza A H1N9 (A/mallard/Ohio/265/1987) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40136V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Ohio/265/1987(H1N9)) hemagglutinin (ABK40634.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0431