Learn More
Sino Biological Influenza A H3N1 (A/swine/Korea/PZ72-1/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40140V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Korea/PZ72-1/2006 (H3N1)) hemagglutinin (ACS71642.1)(Met1-Trp530) , termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0433