Learn More
Sino Biological Influenza A H3N2 (A/Victoria/361/2011) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40145V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/361/2011(H3N2)) hemagglutinin (AGB08328.1)(Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0443